Product Details

SNP ID
rs151251728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:138348126 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCATGATCTGGAAGAAAGACGT[C/T]TCAGGATTCGAAGGGAATGCAGCTA
Phenotype
MIM: 608435
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRAS PubMed Links

Gene Details

Gene
MRAS
Gene Name
muscle RAS oncogene homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085049.2 479 Intron NP_001078518.1
NM_001252090.1 479 Intron NP_001239019.1
NM_001252091.1 479 UTR 5 NP_001239020.1
NM_001252092.1 479 Intron NP_001239021.1
NM_001252093.1 479 Intron NP_001239022.1
NM_012219.4 479 UTR 5 NP_036351.3
XM_005247228.1 479 Intron XP_005247285.1
XM_005247229.2 479 Intron XP_005247286.1
XM_017005887.1 479 Intron XP_016861376.1

View Full Product Details