Product Details

SNP ID
rs137861806
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:185364725 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTAAAAAAATACCCAAAGTTGGCT[A/C]TTGATAAAAATTATCCTTCAAAAGA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LRP2BP PubMed Links

Gene Details

Gene
LRP2BP
Gene Name
LRP2 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018409.3 4641 UTR 3 NP_060879.2
XM_005263125.4 4641 Intron XP_005263182.1
XM_006714260.3 4641 UTR 3 XP_006714323.1
XM_006714265.3 4641 Intron XP_006714328.1
XM_011532110.2 4641 Intron XP_011530412.1
XM_011532111.2 4641 Intron XP_011530413.1
XM_017008410.1 4641 UTR 3 XP_016863899.1
XM_017008411.1 4641 Intron XP_016863900.1
Gene
SNX25
Gene Name
sorting nexin 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317781.1 4641 Intron NP_001304710.1
XM_005263267.4 4641 Intron XP_005263324.1
XM_011532302.2 4641 Intron XP_011530604.1
XM_011532303.2 4641 Intron XP_011530605.1
XM_011532304.2 4641 Intron XP_011530606.1
XM_011532305.2 4641 Intron XP_011530607.1
XM_011532306.2 4641 Intron XP_011530608.1
XM_011532307.2 4641 Intron XP_011530609.1
XM_011532308.2 4641 Intron XP_011530610.1
XM_017008680.1 4641 Intron XP_016864169.1
XM_017008681.1 4641 Intron XP_016864170.1
XM_017008682.1 4641 Intron XP_016864171.1
XM_017008683.1 4641 Intron XP_016864172.1
XM_017008684.1 4641 Intron XP_016864173.1
XM_017008685.1 4641 Intron XP_016864174.1
XM_017008686.1 4641 Intron XP_016864175.1
XM_017008687.1 4641 Intron XP_016864176.1

View Full Product Details