Product Details

SNP ID
rs138330999
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:121761565 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTCCAAGTTTCCACCCCTGCAAG[C/T]GAACCAAAAACAAATCCTGGCACCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PP12613 PubMed Links

Gene Details

Gene
PP12613
Gene Name
uncharacterized LOC100192379
There are no transcripts associated with this gene.

Gene
TMEM155
Gene Name
transmembrane protein 155
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317837.1 722 Missense Mutation NP_001304766.1
NM_001317838.1 722 Missense Mutation NP_001304767.1
NM_001317839.1 722 Missense Mutation NP_001304768.1
NM_001317841.1 722 Missense Mutation NP_001304770.1
NM_001317842.1 722 Missense Mutation NP_001304771.1
NM_152399.3 722 Missense Mutation NP_689612.3
XM_005262737.2 722 Missense Mutation XP_005262794.1
XM_017007725.1 722 Missense Mutation XP_016863214.1
XM_017007726.1 722 Missense Mutation XP_016863215.1

View Full Product Details