Product Details

SNP ID
rs139205806
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:183446322 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGAGATCGATCTGTTTCCAGCC[A/T]AAGCAGCAGCAGCGTTTCCTCTCAG
Phenotype
MIM: 615914
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CDKN2AIP PubMed Links

Gene Details

Gene
CDKN2AIP
Gene Name
CDKN2A interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317343.1 814 UTR 3 NP_001304272.1
NM_017632.3 814 Missense Mutation CAA,CTA Q213L NP_060102.1

View Full Product Details