Product Details

SNP ID
rs139628331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:53464237 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCAATTTATATGAACTATGACC[A/G]TTAACTTAATAGATTCCAGTGAGTA
Phenotype
MIM: 607686 MIM: 609732
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FIP1L1 PubMed Links

Gene Details

Gene
FIP1L1
Gene Name
factor interacting with PAPOLA and CPSF1
There are no transcripts associated with this gene.

Gene
LNX1
Gene Name
ligand of numb-protein X 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126328.2 Intron NP_001119800.1
NM_032622.2 Intron NP_116011.2
XM_005265785.4 Intron XP_005265842.1
XM_017008776.1 Intron XP_016864265.1

View Full Product Details