Product Details

SNP ID
rs139835933
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:151103933 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGCTAAGAGGGTGTAATGGAAAA[A/C]GCATAAGGCTTGGACTCAGAAGACT
Phenotype
MIM: 180478 MIM: 603568
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RPS3A PubMed Links

Gene Details

Gene
RPS3A
Gene Name
ribosomal protein S3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006.4 Intron NP_000997.1
NM_001267699.1 Intron NP_001254628.1
Gene
SNORD73A
Gene Name
small nucleolar RNA, C/D box 73A
There are no transcripts associated with this gene.

View Full Product Details