Product Details

SNP ID
rs140798165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:169737721 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTATCTACTGGAACAACCAAGCC[C/T]GCACCATGAAAGGTCTTTGTCACAA
Phenotype
MIM: 616614
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HPF1 PubMed Links

Gene Details

Gene
HPF1
Gene Name
histone PARylation factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017867.2 752 Intron NP_060337.2
XM_017008332.1 752 Silent Mutation XP_016863821.1

View Full Product Details