Product Details

SNP ID
rs142664782
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:5051888 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGGGCGGGAACCACTCCCACAAG[A/C]CCCCCGTGTTTGACGAGAATGAGGA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC107986255 PubMed Links

Gene Details

Gene
LOC107986255
Gene Name
translation initiation factor IF-2-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017008890.1 372 Intron XP_016864379.1
Gene
STK32B
Gene Name
serine/threonine kinase 32B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306082.1 372 UTR 5 NP_001293011.1
NM_018401.2 372 Missense Mutation ACC,CCC T9P NP_060871.1
XM_005247982.3 372 Intron XP_005248039.1
XM_005247983.4 372 Intron XP_005248040.1
XM_011513497.1 372 Missense Mutation ACC,CCC T9P XP_011511799.1
XM_011513501.2 372 Intron XP_011511803.1
XM_011513504.1 372 Intron XP_011511806.1
XM_017008374.1 372 Intron XP_016863863.1
XM_017008375.1 372 Intron XP_016863864.1
XM_017008376.1 372 Intron XP_016863865.1
XM_017008377.1 372 Intron XP_016863866.1

View Full Product Details