Product Details

SNP ID
rs142734014
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:152632695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAGTTTATCCCTGGGTAGTTTAC[A/G]TTTGTGCCATTTTTATGATATTACC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM154 PubMed Links

Gene Details

Gene
TMEM154
Gene Name
transmembrane protein 154
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152680.2 Intron NP_689893.1
XM_011531716.2 Intron XP_011530018.1

View Full Product Details