Product Details

SNP ID
rs142767113
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:17804035 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCACCATAGAGTCCTCTTCTTCA[C/G]AGGAATCCCACTCTTCCCCAGAACT
Phenotype
MIM: 606280
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DCAF16 PubMed Links

Gene Details

Gene
DCAF16
Gene Name
DDB1 and CUL4 associated factor 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017741.3 1052 Missense Mutation TCT,TGT S36C NP_060211.3
XM_005248169.3 1052 Missense Mutation TCT,TGT S36C XP_005248226.1
XM_005248171.3 1052 Missense Mutation TCT,TGT S36C XP_005248228.1
XM_006713966.3 1052 Missense Mutation TCT,TGT S36C XP_006714029.1
XM_017008327.1 1052 Missense Mutation TCT,TGT S36C XP_016863816.1
XM_017008328.1 1052 Missense Mutation TCT,TGT S36C XP_016863817.1
Gene
NCAPG
Gene Name
non-SMC condensin I complex subunit G
There are no transcripts associated with this gene.

View Full Product Details