Product Details

SNP ID
rs143352342
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:121761624 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGAAAAGCCAGTGCATGGCACAC[A/T]CTGGGTAAATTTTCTCTCTTATTCT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PP12613 PubMed Links

Gene Details

Gene
PP12613
Gene Name
uncharacterized LOC100192379
There are no transcripts associated with this gene.

Gene
TMEM155
Gene Name
transmembrane protein 155
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317837.1 663 Missense Mutation AGA,AGT R42S NP_001304766.1
NM_001317838.1 663 Missense Mutation AGA,AGT R42S NP_001304767.1
NM_001317839.1 663 Missense Mutation AGA,AGT R42S NP_001304768.1
NM_001317841.1 663 Missense Mutation AGA,AGT R16S NP_001304770.1
NM_001317842.1 663 Missense Mutation AGA,AGT R16S NP_001304771.1
NM_152399.3 663 Missense Mutation AGA,AGT R42S NP_689612.3
XM_005262737.2 663 Missense Mutation AGA,AGT R42S XP_005262794.1
XM_017007725.1 663 Missense Mutation AGA,AGT R42S XP_016863214.1
XM_017007726.1 663 Missense Mutation AGA,AGT R16S XP_016863215.1

View Full Product Details