Product Details

SNP ID
rs143545447
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:183446169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGAGCAAGATCACGCAAAAACCT[C/G]TGCCAAGACAGAACGTGCATCAGCT
Phenotype
MIM: 615914
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDKN2AIP PubMed Links

Gene Details

Gene
CDKN2AIP
Gene Name
CDKN2A interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317343.1 661 UTR 3 NP_001304272.1
NM_017632.3 661 Missense Mutation TCT,TGT S162C NP_060102.1

View Full Product Details