Product Details

SNP ID
rs144059414
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:184695390 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTAAATAACAGAGCTGGAAGGCT[C/G]GATGAATCATACTGTTGAAAGAAAA
Phenotype
MIM: 611511 MIM: 615421
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CENPU PubMed Links

Gene Details

Gene
CENPU
Gene Name
centromere protein U
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024629.3 1225 Silent Mutation TCC,TCG S385S NP_078905.2
XM_005263218.4 1225 Silent Mutation TCC,TCG S415S XP_005263275.2
Gene
PRIMPOL
Gene Name
primase and DNA directed polymerase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300767.1 1225 Intron NP_001287696.1
NM_001300768.1 1225 Intron NP_001287697.1
NM_152683.3 1225 Intron NP_689896.1
XM_011531719.1 1225 Intron XP_011530021.1
XM_011531720.1 1225 Intron XP_011530022.1
XM_011531721.2 1225 Intron XP_011530023.1
XM_011531723.1 1225 Intron XP_011530025.1
XM_011531724.2 1225 Intron XP_011530026.1
XM_011531725.1 1225 Intron XP_011530027.1
XM_011531726.2 1225 Intron XP_011530028.1
XM_011531729.1 1225 Intron XP_011530031.1
XM_011531730.2 1225 Intron XP_011530032.1
XM_017007864.1 1225 Intron XP_016863353.1
XM_017007865.1 1225 Intron XP_016863354.1
XM_017007866.1 1225 Intron XP_016863355.1
XM_017007867.1 1225 Intron XP_016863356.1
XM_017007868.1 1225 Intron XP_016863357.1
XM_017007869.1 1225 Intron XP_016863358.1
XM_017007870.1 1225 Intron XP_016863359.1
XM_017007871.1 1225 Intron XP_016863360.1
XM_017007872.1 1225 Intron XP_016863361.1
XM_017007873.1 1225 Intron XP_016863362.1
XM_017007874.1 1225 Intron XP_016863363.1
XM_017007875.1 1225 Intron XP_016863364.1

View Full Product Details