Product Details

SNP ID
rs144869043
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:68478895 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAAGCACAGGTGACTCTCATAGA[C/T]GCTACAACTTGCAATGAACCTCAAG
Phenotype
MIM: 610399
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TMPRSS11E PubMed Links

Gene Details

Gene
TMPRSS11E
Gene Name
transmembrane protease, serine 11E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014058.3 1078 Silent Mutation GAC,GAT D338D NP_054777.2
XM_011531896.2 1078 Silent Mutation GAC,GAT D260D XP_011530198.1

View Full Product Details