Product Details

SNP ID
rs145666261
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:93829728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCGGAGGGAGCCAGCGGCCGACC[C/T]CGCCCGGGAGTTGCCGGACTCGCTT
Phenotype
MIM: 601461
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATOH1 PubMed Links

Gene Details

Gene
ATOH1
Gene Name
atonal bHLH transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005172.1 802 Missense Mutation CCG,TCG P268S NP_005163.1

View Full Product Details