Product Details

SNP ID
rs145768751
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:68476391 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGAGTCATCGCTGTGGAGCAAC[C/T]TTAATTAATGCCACATGGCTTGTGA
Phenotype
MIM: 610399
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMPRSS11E PubMed Links

Gene Details

Gene
TMPRSS11E
Gene Name
transmembrane protease, serine 11E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014058.3 724 Silent Mutation ACC,ACT T220T NP_054777.2
XM_011531896.2 724 Silent Mutation ACC,ACT T142T XP_011530198.1

View Full Product Details