Product Details

SNP ID
rs145801188
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:68466731 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTTCTAACAATTTTACAGAAAT[A/G]AGCCAGAGACTTGAATCAATGGTAA
Phenotype
MIM: 610399
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TMPRSS11E PubMed Links

Gene Details

Gene
TMPRSS11E
Gene Name
transmembrane protease, serine 11E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014058.3 301 Missense Mutation ATA,ATG I79M NP_054777.2
XM_011531896.2 301 Missense Mutation ATA,ATG I1M XP_011530198.1

View Full Product Details