Product Details

SNP ID
rs146368553
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:150079216 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCTACCGCACGCGGACCCTGCA[A/G]GCCCTCAGCTCGGAGAAGAAGGCCA
Phenotype
MIM: 613166
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
DCLK2 PubMed Links

Gene Details

Gene
DCLK2
Gene Name
doublecortin like kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040260.3 943 Silent Mutation CAA,CAG Q63Q NP_001035350.2
NM_001040261.4 943 Silent Mutation CAA,CAG Q63Q NP_001035351.4
XM_005262785.2 943 Silent Mutation CAA,CAG Q63Q XP_005262842.1
XM_005262786.2 943 Silent Mutation CAA,CAG Q63Q XP_005262843.1
XM_017007832.1 943 Silent Mutation CAA,CAG Q63Q XP_016863321.1
XM_017007833.1 943 Silent Mutation CAA,CAG Q63Q XP_016863322.1

View Full Product Details