Product Details
- SNP ID
-
rs146496147
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:121669691 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAATCTCACTCCTGGAAACCATGA[C/T]TCTGATGAGGGTATGATCATCTGTC
- Phenotype
-
MIM: 131230
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANXA5
PubMed Links
Gene Details
- Gene
- ANXA5
- Gene Name
- annexin A5
View Full Product Details