Product Details

SNP ID
rs147620158
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:93829797 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGGGTACTCGGTGCAGCTGGAC[A/G]CTCTGCACTTCTCGACTTTCGAGGA
Phenotype
MIM: 601461
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATOH1 PubMed Links

Gene Details

Gene
ATOH1
Gene Name
atonal bHLH transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005172.1 871 Missense Mutation ACT,GCT T291A NP_005163.1

View Full Product Details