Product Details

SNP ID
rs147662951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:184757226 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGTTGGAGTCAGAAGGCCATTGT[C/T]GATAGAAAATAATTCAGGGTGCAAT
Phenotype
MIM: 152425
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACSL1 PubMed Links

Gene Details

Gene
ACSL1
Gene Name
acyl-CoA synthetase long-chain family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286708.1 2181 Missense Mutation NP_001273637.1
NM_001286710.1 2181 Missense Mutation NP_001273639.1
NM_001286711.1 2181 Missense Mutation NP_001273640.1
NM_001286712.1 2181 Missense Mutation NP_001273641.1
NM_001995.3 2181 Missense Mutation NP_001986.2
XM_005262828.1 2181 Missense Mutation XP_005262885.1
XM_005262829.1 2181 Missense Mutation XP_005262886.1
XM_005262831.1 2181 Missense Mutation XP_005262888.1
XM_011531742.1 2181 Missense Mutation XP_011530044.1
XM_017007887.1 2181 Missense Mutation XP_016863376.1
XM_017007888.1 2181 Missense Mutation XP_016863377.1
XM_017007889.1 2181 Missense Mutation XP_016863378.1

View Full Product Details