Product Details
- SNP ID
-
rs149261175
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:936616 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACAAATTCAGCAGCTTACACTAAA[C/T]GAACAAATTCCTGGAAAGCACAAAC
- Phenotype
-
MIM: 602052
MIM: 616660
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GAK
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs552617763] are located under a probe and SNP(s) [rs552617763] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GAK
- Gene Name
- cyclin G associated kinase
There are no transcripts associated with this gene.
- Gene
- TMEM175
- Gene Name
- transmembrane protein 175
View Full Product Details