Product Details

SNP ID
rs149266206
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:1348119 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATCAGAAACTTTCGAAGTTGGTA[A/G]AAGAGCTCACAACTTCAGGAGAACC
Phenotype
MIM: 606801 MIM: 614632
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAEA PubMed Links

Gene Details

Gene
MAEA
Gene Name
macrophage erythroblast attacher
There are no transcripts associated with this gene.

Gene
UVSSA
Gene Name
UV stimulated scaffold protein A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317934.1 798 Missense Mutation AAA,GAA K10E NP_001304863.1
NM_001317935.1 798 Missense Mutation AAA,GAA K10E NP_001304864.1
NM_020894.3 798 Missense Mutation AAA,GAA K10E NP_065945.2
XM_017008490.1 798 Missense Mutation AAA,GAA K10E XP_016863979.1
XM_017008491.1 798 Missense Mutation AAA,GAA K10E XP_016863980.1
XM_017008492.1 798 Missense Mutation AAA,GAA K10E XP_016863981.1
XM_017008493.1 798 Missense Mutation AAA,GAA K10E XP_016863982.1
XM_017008494.1 798 Missense Mutation AAA,GAA K10E XP_016863983.1
XM_017008495.1 798 Intron XP_016863984.1
XM_017008496.1 798 Missense Mutation AAA,GAA K119E XP_016863985.1
XM_017008497.1 798 Missense Mutation AAA,GAA K119E XP_016863986.1
XM_017008498.1 798 Missense Mutation AAA,GAA K10E XP_016863987.1
XM_017008499.1 798 Missense Mutation AAA,GAA K10E XP_016863988.1
XM_017008500.1 798 Intron XP_016863989.1

View Full Product Details