Product Details

SNP ID
rs150956771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:169737705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTACCCAACATCATTTTTATCTA[C/G]TGGAACAACCAAGCCTGCACCATGA
Phenotype
MIM: 616614
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HPF1 PubMed Links

Gene Details

Gene
HPF1
Gene Name
histone PARylation factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017867.2 768 Intron NP_060337.2
XM_017008332.1 768 Missense Mutation CTA,GTA L231V XP_016863821.1

View Full Product Details