Product Details
- SNP ID
-
rs138619288
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:150896309 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGTTCTCTGATGTACAACAAGATG[A/G]AACTTCTCACTGAAGGCTTTTCCGC
- Phenotype
-
MIM: 608212
MIM: 612429
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
IRGM
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17800771] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IRGM
- Gene Name
- immunity related GTPase M
- Gene
- ZNF300
- Gene Name
- zinc finger protein 300
View Full Product Details