Product Details

SNP ID
rs138794309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:115832623 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACCTCATAGAGAGTTCCAACTTC[C/T]TGGTCTGGGGAAGGAGCAAAGGACT
Phenotype
MIM: 601507 MIM: 609608
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AP3S1 PubMed Links

Gene Details

Gene
AP3S1
Gene Name
adaptor related protein complex 3 sigma 1 subunit
There are no transcripts associated with this gene.

Gene
ATG12
Gene Name
autophagy related 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277783.1 504 Missense Mutation AGA,GGA R69G NP_001264712.1
NM_004707.3 504 Silent Mutation CAA,CAG Q114Q NP_004698.3

View Full Product Details