Product Details

SNP ID
rs139089540
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:10252094 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAGCAGCATTCTCAAGTTTGGCA[C/T]ATTGACATTCGAGGTCGGATCATTC
Phenotype
MIM: 610150
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCT5 PubMed Links

Gene Details

Gene
CCT5
Gene Name
chaperonin containing TCP1 subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306153.1 Intron NP_001293082.1
NM_001306154.1 Intron NP_001293083.1
NM_001306155.1 Intron NP_001293084.1
NM_001306156.1 Intron NP_001293085.1
NM_012073.4 Intron NP_036205.1
Gene
FAM173B
Gene Name
family with sequence similarity 173 member B
There are no transcripts associated with this gene.

View Full Product Details