Product Details

SNP ID
rs139369020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:151259768 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTGTCCCCAGCCATCCCAGCTCA[A/G]TAGCTTTTCCTGGGATAACTGTGAT
Phenotype
MIM: 613109
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GM2A PubMed Links

Gene Details

Gene
GM2A
Gene Name
GM2 ganglioside activator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000405.4 260 Missense Mutation AAT,AGT N32S NP_000396.2
NM_001167607.1 260 Missense Mutation AAT,AGT N32S NP_001161079.1

View Full Product Details