Product Details
- SNP ID
-
rs139629514
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:110739263 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCACTGGGTGACGACTCCCCCAGA[C/T]ATCCCCGGCAGCCGCAACCTGCACT
- Phenotype
-
MIM: 610826
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SLC25A46
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs145421520] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLC25A46
- Gene Name
- solute carrier family 25 member 46
There are no transcripts associated with this gene.
- Gene
- TMEM232
- Gene Name
- transmembrane protein 232
View Full Product Details