Product Details

SNP ID
rs139789028
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:127657554 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGACAGCCCATCCCCTCATCCCT[A/T]GTGCCCCTTGGGAACGAATCAGCAG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CTXN3 PubMed Links

Gene Details

Gene
CTXN3
Gene Name
cortexin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001048252.2 607 Silent Mutation CTA,CTT L11L NP_001041717.1
NM_001127385.1 607 Silent Mutation CTA,CTT L11L NP_001120857.1

View Full Product Details