Product Details

SNP ID
rs140560109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:154477579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGCCAGCACGTCCATCAGGTAGG[C/T]GATGTAGCTGGTGGCTAGGCGCAGA
Phenotype
MIM: 602406
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HAND1 PubMed Links

Gene Details

Gene
HAND1
Gene Name
heart and neural crest derivatives expressed 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004821.2 631 Missense Mutation NP_004812.1
XM_005268531.1 631 Missense Mutation XP_005268588.1

View Full Product Details