Product Details

SNP ID
rs140871446
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:90459804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATGTATATGTTTATGTGTGTGTG[A/G]GTTGAATGAATAAAAAACTAAAGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MBLAC2 PubMed Links

Gene Details

Gene
MBLAC2
Gene Name
metallo-beta-lactamase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203406.1 2679 UTR 3 NP_981951.1

View Full Product Details