Product Details

SNP ID
rs141021686
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:56909652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGCCCCGCGGGGCAGCCATGCCT[A/G]GCCGTCTGCTGCGGGGCCTGTGGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIER3 PubMed Links

Gene Details

Gene
MIER3
Gene Name
MIER family member 3
There are no transcripts associated with this gene.

Gene
SETD9
Gene Name
SET domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171990.2 393 Missense Mutation AGC,GGC S3G NP_001165461.1
NM_001323018.1 393 Intron NP_001309947.1
NM_001323019.1 393 Missense Mutation AGC,GGC S3G NP_001309948.1
NM_001323020.1 393 Intron NP_001309949.1
NM_001323022.1 393 Intron NP_001309951.1
NM_153706.3 393 Missense Mutation AGC,GGC S3G NP_714917.2
XM_005248426.4 393 Intron XP_005248483.1
XM_011543139.2 393 Intron XP_011541441.1
XM_011543140.2 393 Intron XP_011541442.1

View Full Product Details