Product Details

SNP ID
rs141240226
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:87268962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAATACGAGGAGGAAGAGGTGGCC[A/G]TACCGTTGACCGCTCCTCCAACTAA
Phenotype
MIM: 139150
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RASA1 PubMed Links

Gene Details

Gene
RASA1
Gene Name
RAS p21 protein activator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002890.2 1162 Missense Mutation ATA,GTA I171V NP_002881.1
NM_022650.2 1162 UTR 5 NP_072179.1
XM_011543525.1 1162 Missense Mutation ATA,GTA I171V XP_011541827.1
XM_011543527.2 1162 Missense Mutation ATA,GTA I171V XP_011541829.1

View Full Product Details