Product Details

SNP ID
rs141374326
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141150829 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCGCAGCGAAGGCAGGAAGTTCC[C/T]GGAGCTGGTGCTAGACAAACCGTTG
Phenotype
MIM: 606332
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCDHB17P PubMed Links

Gene Details

Gene
PCDHB17P
Gene Name
protocadherin beta 17 pseudogene
There are no transcripts associated with this gene.

Gene
PCDHB6
Gene Name
protocadherin beta 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303145.1 603 Missense Mutation CCG,CTG P55L NP_001290074.1
NM_018939.3 603 Missense Mutation CCG,CTG P191L NP_061762.2

View Full Product Details