Product Details

SNP ID
rs142164982
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:157675115 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTTAATGAATTCCAGTGAAGCAG[C/T]GATGAAAAAAACTTTACCCAAGAGC
Phenotype
MIM: 606698
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C5orf52 PubMed Links

Gene Details

Gene
C5orf52
Gene Name
chromosome 5 open reading frame 52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145132.1 298 Missense Mutation GCG,GTG A79V NP_001138604.1
XM_011534416.1 298 Missense Mutation GCG,GTG A79V XP_011532718.1
XM_017008909.1 298 Intron XP_016864398.1
Gene
SOX30
Gene Name
SRY-box 30
There are no transcripts associated with this gene.

View Full Product Details