Product Details

SNP ID
rs142497899
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141924683 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCCACTTTGCTGGTTCCTGTGC[C/T]TAACCTCGACAGGTAAGATACTGCC
Phenotype
MIM: 615741
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA0141 PubMed Links

Gene Details

Gene
KIAA0141
Gene Name
KIAA0141
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142603.2 291 Missense Mutation CCT,CTT P45L NP_001136075.1
NM_014773.4 291 Missense Mutation CCT,CTT P45L NP_055588.3
XM_005268547.3 291 Missense Mutation CCT,CTT P45L XP_005268604.1
XM_005268548.3 291 Missense Mutation CCT,CTT P45L XP_005268605.1
XM_005268552.2 291 Missense Mutation CCT,CTT P45L XP_005268609.1
XM_006714812.3 291 Intron XP_006714875.1
XM_011537715.1 291 Missense Mutation CCT,CTT P45L XP_011536017.1
XM_011537716.2 291 Missense Mutation CCT,CTT P45L XP_011536018.1

View Full Product Details