Product Details

SNP ID
rs142728816
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141356050 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACGTGACGTATTCTTTCCGGAAA[G/T]TAAGAGACAAAATATCACAGCTATT
Phenotype
MIM: 606288 MIM: 606289 MIM: 606290 MIM: 606291 MIM: 606292 MIM: 606299 MIM: 606300
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PCDHGA1 PubMed Links

Gene Details

Gene
PCDHGA1
Gene Name
protocadherin gamma subfamily A, 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018912.2 1026 Intron NP_061735.1
NM_031993.1 1026 Intron NP_114382.1
Gene
PCDHGA2
Gene Name
protocadherin gamma subfamily A, 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018915.3 1026 Intron NP_061738.1
NM_032009.2 1026 Intron NP_114398.1
Gene
PCDHGA3
Gene Name
protocadherin gamma subfamily A, 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018916.3 1026 Intron NP_061739.2
NM_032011.1 1026 Intron NP_114400.1
Gene
PCDHGA4
Gene Name
protocadherin gamma subfamily A, 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018917.3 1026 Missense Mutation GTA,TTA V315L NP_061740.2
NM_032053.2 1026 Missense Mutation GTA,TTA V315L NP_114442.2
Gene
PCDHGA5
Gene Name
protocadherin gamma subfamily A, 5
There are no transcripts associated with this gene.

Gene
PCDHGB1
Gene Name
protocadherin gamma subfamily B, 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018922.2 1026 Intron NP_061745.1
NM_032095.1 1026 Intron NP_115266.1
Gene
PCDHGB2
Gene Name
protocadherin gamma subfamily B, 2
There are no transcripts associated with this gene.

View Full Product Details