Product Details

SNP ID
rs142875628
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:179112506 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGGAAAATAGGAGCTATTAAAG[C/T]TTTCAAGGAGCAAAAGACAATGTAG
Phenotype
MIM: 604539
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAMTS2 PubMed Links

Gene Details

Gene
ADAMTS2
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014244.4 5099 UTR 3 NP_055059.2
NM_021599.3 5099 Intron NP_067610.1
XM_017010063.1 5099 UTR 3 XP_016865552.1

View Full Product Details