Product Details

SNP ID
rs142930327
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150446830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCTCCTGTGGCCCGGAGTTTGA[C/T]GTGTAGGGCGGTGATACCCAGCTCC
Phenotype
MIM: 130620
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPS14 PubMed Links

Gene Details

Gene
RPS14
Gene Name
ribosomal protein S14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025070.1 556 Missense Mutation ATC,GTC I95V NP_001020241.1
NM_001025071.1 556 Missense Mutation ATC,GTC I95V NP_001020242.1
NM_005617.3 556 Intron NP_005608.1
XM_017009684.1 556 Missense Mutation ATC,GTC I95V XP_016865173.1

View Full Product Details