Product Details

SNP ID
rs142941881
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:90461220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTAGAGCTAAACTTGCAAGAGATC[A/G]CATGGCAAAAGTAGAAACTTTGTGA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MBLAC2 PubMed Links

Gene Details

Gene
MBLAC2
Gene Name
metallo-beta-lactamase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203406.1 1263 Nonsense Mutation CGA,TGA R263* NP_981951.1

View Full Product Details