Product Details

SNP ID
rs143036065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:24487856 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTGAGTCGTAGGGGGGTGCGGTG[C/G]GGTCAAGATCATGCTCTTTTAGCCT
Phenotype
MIM: 604555
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDH10 PubMed Links

Gene Details

Gene
CDH10
Gene Name
cadherin 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317222.1 2177 Missense Mutation CCC,CGC P131R NP_001304151.1
NM_001317224.1 2177 Missense Mutation CCC,CGC P723R NP_001304153.1
NM_006727.4 2177 Missense Mutation CCC,CGC P725R NP_006718.2
XM_011513923.2 2177 Missense Mutation CCC,CGC P725R XP_011512225.1
XM_017008915.1 2177 Missense Mutation CCC,CGC P723R XP_016864404.1

View Full Product Details