Product Details

SNP ID
rs143487813
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:115831203 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGCCATGTGTATTGTGTAAAATT[A/G]CAGTCTTTCAGTCTGTCCTATGTGC
Phenotype
MIM: 609608
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATG12 PubMed Links

Gene Details

Gene
ATG12
Gene Name
autophagy related 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277783.1 1186 UTR 3 NP_001264712.1
NM_004707.3 1186 UTR 3 NP_004698.3

View Full Product Details