Product Details

SNP ID
rs143506945
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:139388127 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCACTTGGTCCACTTCTCGAACTC[C/T]GTAGCTGGTGGCAGTGGAGAGGAGA
Phenotype
MIM: 609447 MIM: 603790
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MZB1 PubMed Links

Gene Details

Gene
MZB1
Gene Name
marginal zone B and B1 cell specific protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016459.3 367 Missense Mutation AGA,GGA R103G NP_057543.2
Gene
PROB1
Gene Name
proline rich basic protein 1
There are no transcripts associated with this gene.

Gene
SLC23A1
Gene Name
solute carrier family 23 member 1
There are no transcripts associated with this gene.

Gene
SPATA24
Gene Name
spermatogenesis associated 24
There are no transcripts associated with this gene.

View Full Product Details