Product Details
- SNP ID
-
rs143605909
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:173317925 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGAAGGTCCCTGAGCCCCAAGGCC[C/T]CCGACTCTGCCTCGGGCATGGGCGT
- Phenotype
-
MIM: 603665
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
STC2
PubMed Links
Gene Details
- Gene
- STC2
- Gene Name
- stanniocalcin 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003714.2 |
2141 |
Silent Mutation |
GGA,GGG |
G277G |
NP_003705.1 |
View Full Product Details