Product Details

SNP ID
rs143954927
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:58493643 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTTGTGCCATTCTCTGGGAACA[C/T]ACAGATGAAATGACCTGAACCCTGC
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GAPT PubMed Links

Gene Details

Gene
GAPT
Gene Name
GRB2-binding adaptor protein, transmembrane
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304428.1 132 Intron NP_001291357.1
NM_001304429.1 132 UTR 5 NP_001291358.1
NM_001304431.1 132 Intron NP_001291360.1
NM_152687.3 132 UTR 5 NP_689900.1

View Full Product Details