Product Details
- SNP ID
-
rs144081956
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:179922114 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTTCATACTCACAAACACTTGAC[A/G]TGGTCAGTCTTGGAAAAGTTTAGGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RNF130
PubMed Links
Gene Details
- Gene
- RNF130
- Gene Name
- ring finger protein 130
View Full Product Details