Product Details

SNP ID
rs144730321
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:124641285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTCATATTGACCCGGGATTAGCG[G/T]GTAACCCATGCCAACGTGGGTGTGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF608 PubMed Links

Gene Details

Gene
ZNF608
Gene Name
zinc finger protein 608
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020747.2 3788 Missense Mutation ACG,CCG T1473P NP_065798.2
XM_005272037.2 3788 Missense Mutation ACG,CCG T1473P XP_005272094.1
XM_005272038.1 3788 Missense Mutation ACG,CCG T1473P XP_005272095.1
XM_005272039.2 3788 Missense Mutation ACG,CCG T1473P XP_005272096.1
XM_005272040.3 3788 Missense Mutation ACG,CCG T1185P XP_005272097.1
XM_011543520.2 3788 Intron XP_011541822.1
XM_011543522.2 3788 Missense Mutation ACG,CCG T1176P XP_011541824.1
XM_011543523.2 3788 Missense Mutation ACG,CCG T1088P XP_011541825.1
XM_017009674.1 3788 Missense Mutation ACG,CCG T1473P XP_016865163.1

View Full Product Details