Product Details
- SNP ID
-
rs144841656
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:72444006 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCATCCTCCTTCTCCTCCTTGCAG[C/G]CTTCCTCCAGCACCTCGGGGGCGTG
- Phenotype
-
MIM: 610159
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF366
PubMed Links
Gene Details
- Gene
- ZNF366
- Gene Name
- zinc finger protein 366
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152625.2 |
2171 |
Missense Mutation |
GCC,GGC |
A662G |
NP_689838.1 |
View Full Product Details