Product Details

SNP ID
rs145232171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:154477877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCGTCAGCCGGGCTCAGCAGCCA[C/G]CTCTGGAAGTAGGGCCTTTCCTGAT
Phenotype
MIM: 602406
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HAND1 PubMed Links

Gene Details

Gene
HAND1
Gene Name
heart and neural crest derivatives expressed 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004821.2 333 Silent Mutation AGC,AGG S44R NP_004812.1
XM_005268531.1 333 Silent Mutation AGC,AGG S44R XP_005268588.1

View Full Product Details